Learn More
Sino Biological Mouse Anti-Human IgG Secondary Antibody, Mouse MAb
Supplier:  Sino Biological 68090MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, reconbinant human IgG.This antibody was purified by Protein A affinity chromatography.
Catalog No. 50-255-2921