Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgG1 Fc (103 Cys/Ser) Recombinant Protein

Catalog No. PIRP88064
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88064 1 mg
PIRP88063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88064 Supplier Invitrogen™ Supplier No. RP88064
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. Reconstituted with sterile deionized water to 0.25 mg/mL. Concentration is measured by UV.

Constant region of immunoglobulin heavy chains. Immunoglobulins, also known as antibodies, are membrane-bound or secreted glycoproteins produced by B lymphocytes. In the recognition phase of humoral immunity, the membrane-bound immunoglobulins serve as receptors which, upon binding of a specific antigen, trigger the clonal expansion and differentiation of B lymphocytes into immunoglobulins-secreting plasma cells. Secreted immunoglobulins mediate the effector phase of humoral immunity, which results in the elimination of bound antigens (PubMed:22158414, PubMed:20176268). The antigen binding site is formed by the variable domain of one heavy chain, together with that of its associated light chain. Thus, each immunoglobulin has two antigen binding sites with remarkable affinity for a particular antigen. The variable domains are assembled by a process called V-(D)-J rearrangement and can then be subjected to somatic hypermutations which, after exposure to antigen and selection, allow affinity maturation for a particular antigen (PubMed:17576170, PubMed:20176268). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01857
Concentration 0.25 mg/mL
For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 3500
Molecular Weight (g/mol) 26.08 kDa
Name Human IgG1 Fc (103 Cys/Ser)
pH Range 7.4
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias IgG1; IgG3; Igh-4; Ighg; IGHG1; IGHG2; IGHG3; IGHG4; Immunoglobulin heavy chain (gamma polypeptide); immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 2 (G2m marker); immunoglobulin heavy constant gamma 3 (G3m marker); immunoglobulin heavy constant gamma 4 (G4m marker); RGD1359539; VH7183
Common Name IGHG1
Gene Symbol IGHG1
Biological Activity 1. Measured by its ability to bind human CD16a-His in a functional ELISA. 2. Measured by its ability to bind human CD16a-AVI-His in a functional ELISA. 3.Measured by its ability to bind human CD16a-His in a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Human IgG1 Fc, amino acids Glu99-Lys330 (Accession # P01857-1) with a 103 Cys/Ser mutation
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Immobilized Recombinant Human IgG1-Fc Protein (103 Cys/Ser) (Cat: 10702-HNAH) at 2μg/mL (100μL/well) can bind Recombinant Human CD64 / FCGR1A Protein (His Tag) (Cat: 10256-H08H), the EC50 is 1.6-4.8 ng/mL
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥95% as determined by SDS-PAGE. ≥95% as determined by SEC-HPLC.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.