Learn More
ACROBiosystems Human IgG1 Fc (C103S, N180A) Protein, His Tag
Catalog No. 502331124
Encompass_Preferred
Human IgG1 Fc (C103S, N180A) Protein, His Tag (IG1-H52H7) is expressed from human 293 cells (HEK293). It contains AA Pro 100 - Lys 330 (Accession # P01857-1 (C103S, N180A)).
Catalog No. 50-233-1124 Supplier ACROBiosystems Supplier No. IG1H52H7200UG
Human IgG1 Fc (C103S, N180A) Protein, His Tag (IG1-H52H7) is expressed from human 293 cells (HEK293). It contains AA Pro 100 - Lys 330 (Accession # P01857-1 (C103S, N180A)).