Learn More
ACROBiosystems Human IgG1 Fc (C103S, D239E, L241E) Protein, Tag Free (MALS verified)
Catalog No. 502331940
Encompass_Preferred
Human IgG1 Fc (C103S, D239E, L241E), Tag Free (IG1-H52H1) is expressed from human 293 cells (HEK293). It contains AA Pro 100 - Lys 330 (Accession # P01857 (C103S, D239E, L241E)).
Catalog No. 50-233-1940 Supplier ACROBiosystems Supplier No. IG1H52H1200UG
Human IgG1 Fc (C103S, D239E, L241E), Tag Free (IG1-H52H1) is expressed from human 293 cells (HEK293). It contains AA Pro 100 - Lys 330 (Accession # P01857 (C103S, D239E, L241E)).