Learn More
ACROBiosystems Human IgG1 Fc 25ug
Catalog No. 502534642
Encompass_Preferred
FITC-Labeled Human IgG1 Fc Protein His Tag (IG1-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (C103S M135Y S137T T139E H316K N317F) (Accession # P01857-1 (C103S M135Y S137T T139E H316K N317F)).
Catalog No. 50-253-4642 Supplier ACROBiosystems Supplier No. IG1HF2H325UG
FITC-Labeled Human IgG1 Fc Protein His Tag (IG1-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (C103S M135Y S137T T139E H316K N317F) (Accession # P01857-1 (C103S M135Y S137T T139E H316K N317F)).