Learn More
Immunology Consultants Human IgG1 Fc - Recombinant
SDP
Catalog No. 5000435328
Encompass_Preferred
This recombinant Human IgG1 Fc fragment offers a dependable, high‑quality antigen for immunoassay development, Fc receptor studies, and antibody engineering workflows. IgG1 is the most abundant and functionally active human IgG subclass, making its Fc region a critical component in therapeutic antibody design and analytical testing. Provided at 1 mg, this Fc fragment supports ELISA calibration, binding assays, and Fc‑specific antibody screening. Recombinant production ensures structural fidelity and consistent performance across applications. A valuable reagent for teams working on IgG1‑based therapeutics, Fc‑mediated effector function studies, or subclass‑specific assay validation.
Catalog No. 50-004-35328 Supplier Immunology Consultants Supplier No. AG80023502
This recombinant Human IgG1 Fc fragment offers a dependable, high‑quality antigen for immunoassay development, Fc receptor studies, and antibody engineering workflows. IgG1 is the most abundant and functionally active human IgG subclass, making its Fc region a critical component in therapeutic antibody design and analytical testing. Provided at 1 mg, this Fc fragment supports ELISA calibration, binding assays, and Fc‑specific antibody screening. Recombinant production ensures structural fidelity and consistent performance across applications. A valuable reagent for teams working on IgG1‑based therapeutics, Fc‑mediated effector function studies, or subclass‑specific assay validation.