Learn More
AdipoGen Human IgG1 Isotype Ctrl
SDP
Supplier:  AdipoGen AG35B0006C100
Encompass_Preferred
Monoclonal Antibody. Isotype Human IgG1. Applications FACS, ICC, IHC, IP. Lyophilized from PBS. Negative control in assays where proteins fused to human Fc IgG1 or human IgG1 monoclonal antibodies are used.
Catalog No. 50-256-3086