Learn More
ANCELL CORPORATION Human IgG1κ Myeloma, High Purity, Ideal Negative Control, 100 µg
Supplier:  ANCELL CORPORATION 295810
Encompass
The Human IgG1 Myeloma is a purified immunoglobulin designed for research applications. With a concentration of 1.0 mg/ml and >95% purity, it serves as a reliable negative control for immunological assays involving IgG1 antibodies, enhancing research specificity. Its human origin and optimized formulation ensure stability and integrity during experiments, making it essential for reliable outcomes.
Catalog No. NC0905765