Learn More
Carbosynth LLC. Purified Human IgG1 protein
SDP
Supplier:  Carbosynth LLC. 31AI171MG
Encompass_Preferred
In normal adult serum, the approximate percentage composition of IgG with respect to its subclasses is IgG1:65%, Ig2:23%, IgG3:6%, and IgG4:6%. Abnormal levels of one or more subclasses may be associated with autoimmune, anaphylatic and gut diseases, as well as with recurrent infection.
Catalog No. 50-125-3279