Learn More
ABclonal Technology Human IgG3 Rabbit mAb
SDP
Catalog No. 5000005019
Encompass_Preferred
Predicted to enable antigen binding activity and immunoglobulin receptor binding activity Involved in retina homeostasis Located in blood microparticle and extracellular exosome IgG is a monomeric immunoglobulin built of two heavy chains gamma and two light chains Each molecule has two antigen binding sites This is the most abundant immunoglobulin and is approximately equally distributed in blood and in tissue liquids constituting 75% of serum immunoglobulins in humans There are 4 subclasses IgG1 (66%) IgG2 (23%) IgG3 (7%) and IgG4 (4%)
Catalog No. 50-000-05019 Supplier ABclonal Technology Supplier No. A23914100UL
Predicted to enable antigen binding activity and immunoglobulin receptor binding activity Involved in retina homeostasis Located in blood microparticle and extracellular exosome IgG is a monomeric immunoglobulin built of two heavy chains gamma and two light chains Each molecule has two antigen binding sites This is the most abundant immunoglobulin and is approximately equally distributed in blood and in tissue liquids constituting 75% of serum immunoglobulins in humans There are 4 subclasses IgG1 (66%) IgG2 (23%) IgG3 (7%) and IgG4 (4%)