Learn More
Sino Biological IgE Fc Protein, Human, Recombinant (His Tag)
Supplier:  Sino Biological 29702H08H11
Encompass_Preferred
A DNA sequence encoding the human IGHE (Cys104-Lys427) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3819