Learn More
Sino Biological Human IGHG1 PAb 20ug (Sino Biological)
Catalog No. 502370533
Encompass_Preferred
Produced in rabbits immunized with recombinant human IgG1-Fc (Catalog#10702-HNAH; P01857-1; Glu99-Lys330,C103S). This polyclonal antibody was purified by antigen affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-237-0533 Supplier Sino Biological Supplier No. 10702T16H20
Produced in rabbits immunized with recombinant human IgG1-Fc (Catalog#10702-HNAH; P01857-1; Glu99-Lys330,C103S). This polyclonal antibody was purified by antigen affinity chromatography and conjugated with horseradish-peroxidase (HRP).