Learn More
Sino Biological Recombinant Human IgG1-Fc Protein (103 Cys/Ser), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10702HNAHB20
Encompass_Preferred
A DNA sequence encoding the Human IGHG1 (AAC82527.1) (Glu99-Lys330) (one aa mutation,103 Cys/Ser) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5195