Learn More
Sino Biological Recombinant Human IgG1-Fc Protein (C103S, AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10702H17HB20
Encompass_Preferred
A DNA sequence encoding the human IGHG1 (AAC82527.1) (Glu99-Lys330) was fused with an AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1506