Learn More
Sino Biological Recombinant Human IgG4 Protein (S108P), HPLC-verified
Supplier:  Sino Biological 13505HNAH1
Encompass_Preferred
A DNA sequence encoding the human IgG4 Fc region (P01861) (Glu 99-Lys 327, with mutation Ser 108 Pro) was expressed and purified.
Catalog No. 50-255-4226