Learn More
Sino Biological Anti-IGSF11 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13094T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IGSF11 (rh IGSF11; Catalog#13094-H08H; AAH34411.1; Met1-Gly240). IGSF11 specific IgG was purified by Human IGSF11 affinity chromatography.
Catalog No. 50-255-4316