Learn More
Sino Biological Anti-IGSF3 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11290MM05P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IGSF3 (rh IGSF3; Catalog#11290-H08H; O75054-1; Met1-Ala1124) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4718