Learn More
Kingfisher Biotech Inc Human IL-1 beta Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0877H025
Encompass_Preferred
The Human IL-1 beta yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human IL-1 beta applications are for cell culture, ELISA standard, and Western Blot Control. The Human IL-1 beta yeast-derived recombinant protein can be purchased in multiple sizes. Human IL-1 beta Specifications: (Molecular Weight: 17.4 kDa) (Amino Acid Sequence: APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS (153)) (Gene ID: 3553). For research use only. Made in the USA
Catalog No. 50-241-3187