Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-1 beta, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF20001B10
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF20001B10 10 μg
AF20001B100 100 μg
AF20001B1MG 1 mg
AF20001B500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF20001B10 Supplier Gibco™ Supplier No. AF20001B10UG
  • $250.00
    $187.65 / Each of 1

    Save $62.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-1beta is a 17.3 kDa protein containing 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01584
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3553
Molecular Weight (g/mol) 17.3 kDa
Name Human IL-1 beta, Animal-Free
Purification Method Purified
Quantity 10 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Common Name IL-1 beta
Gene Symbol IL1B
Biological Activity Determined by its ability to stimulate the proliferation of mouse D10S cells. The expected ED50 is ≤ 0.001 ng/ml, corresponding to a specific activity of ≥ 1 x 10^9 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of mouse D10S cells. The expected ED50 is ≤ 0.001 ng/ml, corresponding to a specific activity of ≥ 1 x 10^9 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.