Learn More
ACROBiosystems ACROBiosystems Human IL-1 beta / IL-1F2 Protein, His Tag (MALS verified)
Catalog No. 502107068
Encompass_Preferred
Recombinant Protein;20UG;Human IL-1 beta, His Tag (ILB-H51H3) is expressed from E.coli cells. It contains AA Ala 117 - Ser 269 (Accession # P01584-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 19-20 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human IL-1 beta, His Tag (Cat. No. ILB-H51H3) at 10 μg/mL (100 μL/well) can bind Human IL-1 RII, Fc Tag (Cat. No. IL2-H4256) with a linear range of 2-13 ng/mL (QC tested).
Catalog No. 50-210-7068 Supplier ACROBiosystems Supplier No. ILBH51H320UG
Recombinant Protein;20UG;Human IL-1 beta, His Tag (ILB-H51H3) is expressed from E.coli cells. It contains AA Ala 117 - Ser 269 (Accession # P01584-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 19-20 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human IL-1 beta, His Tag (Cat. No. ILB-H51H3) at 10 μg/mL (100 μL/well) can bind Human IL-1 RII, Fc Tag (Cat. No. IL2-H4256) with a linear range of 2-13 ng/mL (QC tested).