Learn More
ACROBiosystems ACROBiosystems Human IL-1 beta / IL-1F2 Protein, Fc Tag (MALS verified)
Catalog No. 502106245
Encompass_Preferred
Recombinant Protein;20UG;Human IL-1 beta, Fc Tag (ILA-H525c) is expressed from human 293 cells (HEK293). It contains AA Ala 117 - Ser 269 (Accession # P01584-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 43.8 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1 RII, His Tag (Cat. No. IL2-H4226) at 5 μg/mL (100 μL/well) can bind Human IL-1 beta, Fc Tag (Cat. No. ILA-H525c) with a linear range of 4.9-39 ng/mL (QC tested).
Catalog No. 50-210-6245 Supplier ACROBiosystems Supplier No. ILAH525C20UG
Recombinant Protein;20UG;Human IL-1 beta, Fc Tag (ILA-H525c) is expressed from human 293 cells (HEK293). It contains AA Ala 117 - Ser 269 (Accession # P01584-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 43.8 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1 RII, His Tag (Cat. No. IL2-H4226) at 5 μg/mL (100 μL/well) can bind Human IL-1 beta, Fc Tag (Cat. No. ILA-H525c) with a linear range of 4.9-39 ng/mL (QC tested).