Learn More
Raybiotech Inc Human IL-1 R6 (IL-1 Rrp2) ELISA Kit
Supplier:  Raybiotech Inc ELHIL1R61
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 24 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human 2B4, ADAM-9, ANG-2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGF R, IFN alpha/beta R2, IGF-II, IGF-II R, IL-24, IL-33, Kallikrein 14, Legumain, LOX-1, MBL, Neprilysin, Notch-1, NOV, Osteoactivin, PD-1, PGRP-5, Serpin A4, sFRP-3, Thrombomodulin, TLR2, TRAIL R1, Transferrin, WIF-1.
Catalog No. 50-194-9424