Learn More
ACROBiosystems Human IL-1 Rrp2 / IL-1 R6 (C154S, C262S) Protein, His Tag (MALS verified)
Catalog No. 502054732
Encompass_Preferred
Recombinant Protein;1MG;Human IL-1 Rrp2 (C154S, C262S), His Tag (IL2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # Q9HB29-1 (C154S, C262S)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1 Rrp2 (C154S, C262S), His Tag (Cat. No. IL2-H52H4) at 1 μg/mL (100 μL/well) can bind Anti-Human IL-1 Rrp2 MAb, Human IgG with a linear range of 0.2-3 ng/mL (Routinely tested).
Catalog No. 50-205-4732 Supplier ACROBiosystems Supplier No. IL2H52H41MG
Recombinant Protein;1MG;Human IL-1 Rrp2 (C154S, C262S), His Tag (IL2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # Q9HB29-1 (C154S, C262S)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1 Rrp2 (C154S, C262S), His Tag (Cat. No. IL2-H52H4) at 1 μg/mL (100 μL/well) can bind Anti-Human IL-1 Rrp2 MAb, Human IgG with a linear range of 0.2-3 ng/mL (Routinely tested).