Learn More
ACROBiosystems ACROBiosystems Human IL-1 Rrp2 / IL-1 R6 Protein, His Tag
Catalog No. 502106669
Encompass_Preferred
Recombinant Protein;100UG;Human IL-1 Rrp2, His Tag (IL2-H52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # Q9HB29-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.8 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6669 Supplier ACROBiosystems Supplier No. IL2H52H6100UG
Recombinant Protein;100UG;Human IL-1 Rrp2, His Tag (IL2-H52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # Q9HB29-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.8 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.