Learn More
FUJIFILM BIOSCIENCES INC Human IL-10, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 100831MG
Encompass_Preferred
Interleukin 10 (IL-10) is an anti-inflammatory cytokine produced by macrophages and type 2 T helper (Th2) cells. IL-10 inhibits the production of pro-inflammatory cytokines such as interferon gamma (IFN-ɣ), tumor necrosis factor alpha (TNF-A), interleukin 2 (IL-2), interleukin 3 (IL-3), interleukin 4 (IL-4), and granulocyte-macrophage colony-stimulating factor (GM-CSF), made by macrophages and regulatory T cells. IL-10 also suppresses antigen presentation on antigen presenting cells, and enhances the survival, proliferation, and antibody production of B cells. Human IL-10 can act on murine cells, while murine IL-10 is not active on human cells.
Catalog No. 50-990-775
Add to Cart
Request a Quote