Learn More
ACROBiosystems ActiveMax Human IL-11 Protein, His Tag
Supplier:  ACROBiosystems IL1H52431MG
Encompass_Preferred
ActiveMax Human IL-11, His Tag (IL1-H5243) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Leu 199 (Accession # P20809-1).
Catalog No. 50-233-1324