Learn More
ACROBiosystems Human IL-11 Protein, His Tag
Supplier: ACROBiosystems IL1H524320UG
Recombinant Protein;20UG;Human IL-11, His Tag (IL1-H5243) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Leu 199 (Accession # P20809-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.0 kDa.