Learn More
ACROBiosystems Human IL-11 R alpha Protein, Fc Tag
Supplier:  ACROBiosystems ILRH5256100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IL-11 R alpha, Fc Tag (ILR-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ala 370 (Accession # Q14626-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.2 kDa.
Catalog No. 50-205-4770