Learn More
ACROBiosystems Human IL-11 R alpha Protein, Fc Tag
Catalog No. 502054769
Encompass_Preferred
Recombinant Protein;1MG;Human IL-11 R alpha, Fc Tag (ILR-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ala 370 (Accession # Q14626-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.2 kDa.
Catalog No. 50-205-4769 Supplier ACROBiosystems Supplier No. ILRH52561MG
Recombinant Protein;1MG;Human IL-11 R alpha, Fc Tag (ILR-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ala 370 (Accession # Q14626-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.2 kDa.