Learn More
Sino Biological Human IL-12 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785186
Encompass_Preferred
The Human IL-12 (IL12A & IL12B Heterodimer) ELISA Pair Set is for the quantitative determination of Human IL-12 (IL12A & IL12B Heterodimer).The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human IL-12 (IL12A & IL12B Heterodimer) coated on a 96-well plate. Standards and samples are added to the wells, and any Human IL-12 (IL12A & IL12B Heterodimer) present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human IL-12 (IL12A & IL12B Heterodimer) monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human IL-12 (IL12A & IL12B Heterodimer) present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85186 Supplier Sino Biological Supplier No. SEKCT01115
The Human IL-12 (IL12A & IL12B Heterodimer) ELISA Pair Set is for the quantitative determination of Human IL-12 (IL12A & IL12B Heterodimer).The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human IL-12 (IL12A & IL12B Heterodimer) coated on a 96-well plate. Standards and samples are added to the wells, and any Human IL-12 (IL12A & IL12B Heterodimer) present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human IL-12 (IL12A & IL12B Heterodimer) monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human IL-12 (IL12A & IL12B Heterodimer) present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.