Learn More
Sino Biological Human IL-12 p40/IL12B Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786152
Encompass_Preferred
The Human IL12B / IL-12B ELISA Pair Set is for the quantitative determination of Human IL12B / IL-12B.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human IL12B / IL-12B coated on a 96-well plate. Standards and samples are added to the wells, and any Human IL12B / IL-12B present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human IL12B / IL-12B monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human IL12B / IL-12B present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86152 Supplier Sino Biological Supplier No. SEKB100525
The Human IL12B / IL-12B ELISA Pair Set is for the quantitative determination of Human IL12B / IL-12B.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human IL12B / IL-12B coated on a 96-well plate. Standards and samples are added to the wells, and any Human IL12B / IL-12B present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human IL12B / IL-12B monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human IL12B / IL-12B present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.