Learn More
ACROBiosystems Human IL-12 R beta 2 200ug
Catalog No. 502534587
Encompass_Preferred
Biotinylated Human IL-12Rbeta2 Protein HisAvitag (ILB-H82E4) is expressed from human 293 cells (HEK293). It contains AA Lys 24 - Asn 622 (Accession # Q99665-1).
Catalog No. 50-253-4587 Supplier ACROBiosystems Supplier No. ILBH82E4200UG
Biotinylated Human IL-12Rbeta2 Protein HisAvitag (ILB-H82E4) is expressed from human 293 cells (HEK293). It contains AA Lys 24 - Asn 622 (Accession # Q99665-1).