Learn More
ACROBiosystems ACROBiosystems Human IL-12B / NKSF2 / p40 Protein, Fc Tag
Supplier:  ACROBiosystems NK2H525850UG
Encompass_Preferred
Recombinant Protein;50UG;Human IL-12B, Fc Tag (NK2-H5258) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (Accession # AAH67499).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 61.3 kDa. The protein migrates as 66-76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-12 R beta 1, His Tag (Cat. No. ILB-H52H9) at 2 μg/mL (100 μL/well) can bind Human IL-12B, Fc Tag (Cat. No. NK2-H5258) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-210-6727