Learn More
FUJIFILM BIOSCIENCES INC Human IL-13, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1008510UG
Encompass_Preferred
Interleukin 13 (IL-13) is a cytokine secreted from type 2 T helper (Th2) cells.  IL-13 has overlapping functions with interleukin 4 (IL-4), including the induction of immunoglobulin E (IgE) secretion from B cells, and the inhibition of interleukin 1 beta (IL-1B), tumor necrosis factor alpha (TNF-A), interleukin 8 (IL-8), and interleukin 6 (IL-6) inflammatory cytokine expression.  IL-13 also regulates immune cell inflammation in response to the pathophysiological changes of surrounding non-immune cells.  The IL-13 receptor consists of the IL-4Ra and IL-13Ra1 subunits.  IL-13 can also bind the IL-13Ra2 receptor with high affinity.  IL-13 functions are mediated through the JAK/STAT signaling pathway.  Human and mouse IL-13 are cross-reactive.  
Catalog No. 50-990-777