Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-13 R alpha 1/IL-13 R1 ELISA Kit

Catalog No. EH257RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH257RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH257RBX10 Supplier Invitrogen™ Supplier No. EH257RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human IL-13 R alpha 1/IL-13 R1 quantitates human IL-13 R alpha 1/IL-13 R1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-13 R alpha 1/IL-13 R1.

The protein encoded by this gene is a subunit of the interleukin 13 receptor. This subunit forms a receptor complex with IL4 receptor alpha, a subunit shared by IL13 and IL4 receptors. This subunit serves as a primary IL13-binding subunit of the IL13 receptor, and may also be a component of IL4 receptors. This protein has been shown to bind tyrosine kinase TYK2, and thus may mediate the signaling processes that lead to the activation of JAK1, STAT3 and STAT6 induced by IL13 and IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78552
Assay Range 0.05-10 ng/mL
Assay Sensitivity 0.042 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI882074; Cancer/testis antigen 19; CD antigen CD213a1; CD213a1; CD213a1 antigen; CT19; IL13 receptor alpha-1 chain; IL-13 receptor subunit alpha-1; IL13R; IL-13R subunit alpha-1; IL-13r[a]; IL13RA; IL-13Ra; IL13RA1; IL-13RA1; IL-13R-alpha-1; interleukin 13 receptor subunit alpha 1; interleukin 13 receptor, alpha 1; interleukin 13 receptor, alpha 1-like; interleukin-13 receptor subunit alpha-1; Interleukin-13-binding protein; LOC100360218; Novel cytokine receptor 4; NR4AI882074; Cancer/testis antigen 19; CD antigen CD213a1; CD213a1; CD213a1 antigen; CT19; IL13 receptor alpha-1 chain; IL-13 receptor subunit alpha-1; IL13R; IL-13R subunit alpha-1; IL-13r[a]; IL13RA; IL-13Ra; IL13RA1; IL-13RA1; IL-13R-alpha-1; interleukin 13 receptor subunit alpha 1; interleukin 13 receptor, alpha 1; interleukin 13 receptor, alpha 1-like; interleukin-13 receptor subunit alpha-1; Interleukin-13-binding protein; LOC100360218; Novel cytokine receptor 4; NR4;
Gene ID (Entrez) 3597
Gene Symbol IL13RA1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL13RA1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.