Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-13 Recombinant Protein

Catalog No. PIRP88069
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88069 100 μg
PIRP88068 5 μg
PIRP88070 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIRP88069 Supplier Invitrogen™ Supplier No. RP88069
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. Reconstitute with sterile deionized water to 0.1 mg/mL. Concentration is measured by UV.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35225
Concentration 0.25 mg/mL
For Use With (Application) Control, Bioactivity
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 3596
Molecular Weight (g/mol) 12.35 kDa
Name Human IL-13
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <0.01 EU/μg
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Common Name IL-13
Gene Symbol Il13
Biological Activity Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323.). The ED50 for this effect is typically 0.5-3 ng/mL
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Human IL13, amino acids Met1-Asn132 (Accession # AAK53823.1)
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323.). The ED50 for this effect is typically 0.5-3 ng/mL.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥95% as determined by SDS-PAGE. ≥95% as determined by SEC-HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.