Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        R&D Systems™ Recombinant Human IL-13 Protein
    
    
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                10 μg
                            
                        
                            
                                100 μg
                            
                        
                            
                                25 μg
                            
                        
                    Description
The Recombinant Human IL-13 Protein is derived from E. coli. The Recombinant Human IL-13 Protein has been validated for the following applications: Bioactivity.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                            This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 
| Gene ID (Entrez) | 3596 | 
| Molecular Weight (g/mol) | 12.3 kDa | 
| Name | IL-13 | 
| Quantity | 25 μg | 
| Source | E. coli-derived human IL-13 protein Gly21-Asn132 | 
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | 
| Conjugate | Unconjugated | 
| Recombinant | Recombinant | 
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction