Learn More
Raybiotech Inc HUMAN IL-15 ELISA
Supplier:  Raybiotech Inc ELHIL151
Encompass_Preferred
RayBio Human IL-15 ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 3 pg/ml - 800 pg/ml
Catalog No. 50-104-3934