Learn More
PBL Assay Science Human Interleukin-15 (IL-15) / soluble IL-15 Receptor Alpha (sIL-15Ra) Heterodimer, Human Cell-Expressed (HCE) [10 micrograms (ug)]
Supplier:  PBL Assay Science 117021
Encompass_Preferred
Recombinant protein Human Interleukin-15 / soluble IL-15Ra heterodimer, Human Cell-Expressed is manufactured by PBL Assay Science (Catalog # 11702-1) for research use only (RUO) and packaged in 10 micrograms (ug). Hu IL-15/sIL-15Ra is expressed in HEK293 cells.
Catalog No. NC1892110