Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-16 (121 aa) Recombinant Protein, PeproTech®

Catalog No. 20016A10UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20016A10UG 10 μg
20016A100UG 100 μg
200-16A-1MG 1 mg
20016A250UG 250 μg
200-16A-2UG 2 μg
20016A500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 20016A10UG Supplier Gibco™ Supplier No. 20016A10UG
  • $225.00
    $168.65 / Each of 1

    Save $56.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-16 is a 12.4 kDa protein consisting of 121 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14005
For Use With (Application) Bioactivity, Control
Formulation Protein with no preservative
Gene ID (Entrez) 3603
Molecular Weight (g/mol) 12.4 kDa
Name Human IL-16 (121 aa)
Purification Method Purified
Quantity 10 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16
Common Name IL-16
Gene Symbol IL16
Biological Activity Not available
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence SAASASAASD VSVESTAEAT VCTVTLEKMS AGLGFSLEGG KGSLHGDKPL TINRIFKGAA SEQSETVQPG DEILQLGGTA MQGLTRFEAW NIIKALPDGP VTIVIRRKSL QSKETTAAGD S
Content And Storage -20°C
Activity Not available
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.