Learn More
FUJIFILM BIOSCIENCES INC Human IL-16, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 1001441MG
Encompass_Preferred
Interleukin 16 (IL-16) is produced by CD4+ and CD8+ T cells and functions as a chemoattractant forlymphocytes, monocytes, eosinophils, dendritic cells, and Langerhans cells. Additionally, IL-16 upregulates the expressionof the interleukin 2 receptor (CD25), modulates T lymphocyte growth, and suppresses the replication of the humanimmunodeficiency virus (HIV) and simian immunodeficiency viruses (SIV).
Catalog No. 50-990-680
Add to Cart
Request a Quote