Learn More
ACROBiosystems Human IL-17 RA & IL-17 RC Heterodimer Protein, Fc Tag&Fc Tag
Catalog No. 502055155
Encompass_Preferred
Recombinant Protein;1MG;Human IL17RA & IL17RC Protein, Fc Tag&Fc Tag (ILC-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (IL-17RA) & Leu 21 - His 465 (IL-17RC) (Accession # Q96F46-1 (IL-17RA) & NP_703190.1 (IL-17RC)).;Human IL17RA & IL17RC Protein, Fc Tag&Fc Tag is produced by co-expression of IL-17RA and IL-17RC, has a calculated MW of 60.0 kDa (IL-17RA) & 75.7 kDa (IL-17RC). Subunit IL-17RA is fused with a human IgG1 Fc tag at the C-terminus and subunit IL-17RC is fused with a human IgG1 Fc tag at the C-terminus. The reducing (R) heterodimer protein migrates as 80-90 kDa and 100-110 kDa due to glycosylation.;Immobilized Human IL-17A&IL-17F Heterodimer Protein, His Tag&Strep II Tag (Cat. No. ILF-H52WF) at 2 μg/mL (100 μL/well) can bind Human IL17RA & IL17RC Protein, Fc Tag&Fc Tag (Cat. No. ILC-H5257) with a linear range of 1-20 ng/mL (QC tested).;Immobilized Human IL17A, Tag Free (Cat. No. ILA-H5118) at 2 μg/mL (100 μL/well) can bind Human IL17RA &
Catalog No. 50-205-5155 Supplier ACROBiosystems Supplier No. ILCH52571MG
Recombinant Protein;1MG;Human IL17RA & IL17RC Protein, Fc Tag&Fc Tag (ILC-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (IL-17RA) & Leu 21 - His 465 (IL-17RC) (Accession # Q96F46-1 (IL-17RA) & NP_703190.1 (IL-17RC)).;Human IL17RA & IL17RC Protein, Fc Tag&Fc Tag is produced by co-expression of IL-17RA and IL-17RC, has a calculated MW of 60.0 kDa (IL-17RA) & 75.7 kDa (IL-17RC). Subunit IL-17RA is fused with a human IgG1 Fc tag at the C-terminus and subunit IL-17RC is fused with a human IgG1 Fc tag at the C-terminus. The reducing (R) heterodimer protein migrates as 80-90 kDa and 100-110 kDa due to glycosylation.;Immobilized Human IL-17A&IL-17F Heterodimer Protein, His Tag&Strep II Tag (Cat. No. ILF-H52WF) at 2 μg/mL (100 μL/well) can bind Human IL17RA & IL17RC Protein, Fc Tag&Fc Tag (Cat. No. ILC-H5257) with a linear range of 1-20 ng/mL (QC tested).;Immobilized Human IL17A, Tag Free (Cat. No. ILA-H5118) at 2 μg/mL (100 μL/well) can bind Human IL17RA &