Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-17 RB ELISA Kit

Catalog No. p-7219453
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH261RBX5 5 x 96 Tests
EH261RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH261RBX5 Supplier Invitrogen™ Supplier No. EH261RBX5
Only null left
Add to Cart
Add to Cart

ELISA

Human IL-17 RB quantitates human IL-17 RB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-17 RB.

Recently, a number of cytokines belonging to the interleukin (IL)-17 family have been identified. These are termed as IL-17B, IL-17C (1) and IL-17E. IL-17 is a potent proinflammatory cytokine that plays roles in a number of diseases including rheumatoid arthritis, multiple sclerosis, and promotion of tumor growth. IL-17B, C, and E like IL-17 are able to induce proinflammatory responses. However, they do not bind to the IL-17 receptor suggesting that additional IL-17R related receptor might exist. Receptor for IL-17B and IL-17E has been independently isolated and has been designated as EV127 (in mouse) and IL-17Rh1 (in human), respectively. IL-17E induces activation of NF-kB pathway and like IL-17 also induces production of IL-8. IL-17Rh1 mRNA is highly expressed in Kidney and liver and at moderate level in small intestine, colon, and brain.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CRL4; cytokine receptor CRL4; cytokine receptor-like 4; ecotropic viral integration site 27; Evi27; IL-17 receptor B; IL-17 receptor homolog 1; IL-17 B; IL-17 B receptor; IL17BR; IL-17 ER; IL17RB; IL-17 RB; IL17RH1; IL-17 Rh1; IL-20; interleukin 17 receptor B; interleukin 17 receptor homolog 1; interleukin 17 B receptor; Interleukin-17 receptor B; interleukin-17 B receptor; MGC138900; MGC138901; MGC5245; UNQ2501/PRO19612; ZCYTO7
Assay Range 0.85 to 200 ng/mL
Assay Sensitivity 0.85 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 55540
Gene Symbol IL17RB
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL17RB
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.