Learn More
ACROBiosystems ACROBiosystems Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag (MALS verified)
Catalog No. 502107247
Encompass_Preferred
Recombinant Protein;1MG;Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag (ILF-H52W6) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) & Arg 31 - Gln 163 (IL-17F) (Accession # Q16552-1 (IL-17A) & Q96PD4-1 (IL-17F)). Two sequential affinity purification steps were used to ensure exact 1:1 molar ratio of IL17A&IL17F heterodimer.;Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag, produced by co-expression of IL-17A and IL-17F, has a calculated MW of 18.5 kDa (IL-17A) and 16.8 kDa (IL-17F). Subunit IL-17A is fused with a Twin Strep tag at the N-terminus and subunit IL-17F is fused with a polyhistidine tag at the N-terminus. The reducing (R) protein migrates as 19-27 kDa due to glycosylation.;Immobilized Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag (Cat. No. ILF-H52W6) at 2 μg/mL (100 μL/well) can bind Human IL-17 RA, Fc Tag (MALS verified) (Cat. No. ILA-H5257) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized
Catalog No. 50-210-7247 Supplier ACROBiosystems Supplier No. ILFH52W61MG
Recombinant Protein;1MG;Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag (ILF-H52W6) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) & Arg 31 - Gln 163 (IL-17F) (Accession # Q16552-1 (IL-17A) & Q96PD4-1 (IL-17F)). Two sequential affinity purification steps were used to ensure exact 1:1 molar ratio of IL17A&IL17F heterodimer.;Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag, produced by co-expression of IL-17A and IL-17F, has a calculated MW of 18.5 kDa (IL-17A) and 16.8 kDa (IL-17F). Subunit IL-17A is fused with a Twin Strep tag at the N-terminus and subunit IL-17F is fused with a polyhistidine tag at the N-terminus. The reducing (R) protein migrates as 19-27 kDa due to glycosylation.;Immobilized Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag (Cat. No. ILF-H52W6) at 2 μg/mL (100 μL/well) can bind Human IL-17 RA, Fc Tag (MALS verified) (Cat. No. ILA-H5257) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized