Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-17A ProQuantum Immunoassay Kit
GREENER_CHOICE

Catalog No. A35611
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A35611 96 Tests
A356115 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A35611 Supplier Invitrogen™ Supplier No. A35611
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16552
Assay Range 0.32-25000 pg/mL
Assay Sensitivity <0.3 pg/mL
Conjugate FAM
Detection Method Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) Protein Quantitation
For Use With (Equipment) qPCR Instruments
Gene Alias Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-ACtla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A;
Gene ID (Entrez) 3605
Gene Symbol IL17A
Incubation Time 1 hr.
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Shipping Condition Dry Ice
Storage Requirements -20°C
Target IL-17A
Test Time 30 min
Total Assay Time 2 hr
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.