Learn More
FUJIFILM BIOSCIENCES INC Human IL-17A (CTLA-8) , Recombinant Protein, Shenandoah Biotechnology, 25 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1008725UG
Encompass_Preferred
Interleukin 17A (IL-17A), also known as CTLA-8, is a member of the IL-17 family of proteins.  IL-17A is a proinflammatory cytokine that is secreted by activated CD4+ and CD8+ T lymphocytes.  IL-17A acts through its receptor, IL-17R, to promote increased cytokine and chemokine secretion.  In turn, the cytokines and chemokines mediate the immunoregulatory function of IL-17A by promoting the proliferation, maturation, and chemoattraction of neutrophils to inflammatory sites.  Elevated levels of IL-17A are associated with rheumatoid arthritis, airway inflammation, allograft rejection, inflammatory bowel disease, psoriasis, cancer, and multiple sclerosis.  Human, mouse, and rat IL-17A show activity on mouse cells.  
Catalog No. 50-900-474