Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-17A Recombinant Protein, PeproTech®

Catalog No. 20017100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20017100UG 100 μg
200-17-1MG 1 mg
20017250UG 250 μg
200-17-25UG 25 μg
20017500UG 500 μg
200-17-50UG 50 μg
200-17-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 20017100UG Supplier Gibco™ Supplier No. 20017100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-17A is a 31.3 kDa, disulfide-linked homodimer of two 137 amino acid, polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16552
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3605
Molecular Weight (g/mol) 31.3 kDa
Name Human IL-17A
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A
Common Name IL-17A
Gene Symbol IL17A
Biological Activity Measured by its ability to induce GROalpha production by HT-29 cells, using a concentration range of 0.5-5.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVA
Content And Storage -20°C
Activity Measured by its ability to induce GROalpha production by HT-29 cells, using a concentration range of 0.5-5.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.