Learn More
Abcam Human IL-17AF ELISA kit, 1 x 96 Tests
Supplier:  Abcam AB2731621X96TESTS
Encompass_Preferred
Human IL-17AF ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-17AF in Human in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2682018