Learn More
ACROBiosystems Biotinylated Human IL-17B Protein, Fc,Avitag™ (MALS verified)
Catalog No. 502331703
Encompass_Preferred
Biotinylated Human IL-17B, Fc,Avitag (ILB-H82F1) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Phe 180 (Accession # Q9UHF5-1).
Catalog No. 50-233-1703 Supplier ACROBiosystems Supplier No. ILBH82F125UG
Biotinylated Human IL-17B, Fc,Avitag (ILB-H82F1) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Phe 180 (Accession # Q9UHF5-1).