Learn More
FUJIFILM Biosciences Human IL-17E (IL-25) , Recombinant Protein, Shenandoah Biotechnology, 25 ug vial
Catalog No. 50900502
Encompass_Preferred
Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-KB).
Catalog No. 50-900-502 Supplier FUJIFILM Biosciences Supplier No. 10014925UG
Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-KB).